SNAP i.d. 2.0 Protein Detection Systems Mini
Catalog # MILFTISNAP2MINI
Supplier: Merck
Specifications
- Detektor:Chemiluminescence fluorescence chromogenic
- Beskrivning:SNAP i.d. 2.0 Protein detection system
- Seq:02
- Förp.:1
Specifications
About this item
Developed to meet the needs of our Western blotting customers, the SNAP i.d. 2.0 system produces blots of a very high quality. Unique vacuum-driven technology and a built-in flow distributor actively drive reagents through the membrane.
Ordering information: This system includes the SNAP i.d. 2.0™ base, two mini blot holding frames, two antibody collection trays, vacuum tubing, rolling pad and blot roller, two wetting trays and a quick start guide.