Polytron® PTG 36/4 Generators
Catalog # KINE9810065
Supplier: KINEMATICA
New Product
Specifications
- Hastighet:34 m/s
- Volym:100 - 2500 ml
- Förp.:1
- Stator/rotor Ø:36 mm
Specifications
About this item
Standard generator rotor/stator with two teeth rows each.
- Making fine emulsions and suspensions
- Mixes and reduces highly intensive solids into fluids
- Used for gaseous solutions and extraktions
- Separates fibres and cellular material into very small particles