Environmental Express® HotBlock® 200 Digestion Systems
Leverantör: Cole Parmer
New Product
Certifikat
Om denna produkt
Easy-to-use external controllers feature automatic shut-off, alarms,and ability to save profi les with ramp and soak settings. Order heating block and control.
- Eliminate corrosion and sample contamination made of nonmetallic materials
- Easily transport samples with included polycarbonate transfer racks
- Mount external controller safely outside the hood
Specifikationer
- Catalog No:
- STUA53206-00
- STUA53206-03
- Mått:
- 15,2×17,8
- 54,6×38,1
- Effekt:
- 240 VAC
- Storlek:
- 15 ml