Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Phosphomolybdotungstic reagent dilute
Phosphomolybdotungstic reagent dilute Reag. Ph. Eur. 1065001
Phosphomolybdotungstic reagent dilute Reag. Ph. Eur. 1065001
Phosphomolybdotungstic reagent dilute Reag. Ph. Eur. 1065001
Catalog # 87894.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Expand image
Phosphomolybdotungstic reagent dilute Reag. Ph. Eur. 1065001
Catalog # 87894.180
Supplier:
VWR Chemicals
CAS Number:
SDS
Certificates
Specifications
Förpackningstyp:
Plastic bottle
Förp.:
100 mL
Seq:
2440
Specifications
About this item
Product Details
Documentation
:
UN:
3264
ADR:
8,III
Product Details
Documentation