Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sudan IV
Sudan IV, Sigma-Aldrich®
Sudan IV, Sigma-Aldrich®
S4261-25G
:
SIGMA ALDRICH MICROSCOPY
:
Sudan IV, Sigma-Aldrich®
S4261-25G
:
SIGMA ALDRICH MICROSCOPY
:
Förpackningstyp:
Glass bottle for solids
Förp.:
25 g
Seq:
3308
:
:
380.45 g/mol
:
12,08538
:
188 °C
:
M-Clarity™ quality level = MQ200
:
201-635-8
:
GHS07
:
Omgivande
:
85-83-6
:
C₂₄H₂₀N₄O
:
MFCD00003893