Thermo Scientific™ Teleshake Magnetic Shaking System
Catalog # HERA50134882
Supplier: Thermo Scientific
Specifications
- Beskrivning:Teleshake magnetic shaking system without controller
- Förp.:1
Specifications
About this item
Conduct powerful shaking applications from 100 to 2000rpm with the Thermo Scientific™ Teleshake magnetic shaking system, a remote-controlled, wear-free magnetic shaking system for integrated shaking applications; suited for 96 and 384 well microtiter plates.
- Easy integration in robotic systems and platform due to small footprint
- Simultaneous control of up to 14 shakers via RS232 interface
- Powerful shaking movements from 100 to 2000 min⁻¹ (rpm), with an amplitude of 2 mm
- Rapid switching between circular and linear agitation
- Minimum of moved masses, therefore little transference of vibrations onto the installation area
- Integrated threads and drilled holes for assembling on robot workbenches