Fold Down Mini Shelf
Fold down mini shelf - solid, 1200×450×260 mm
Katalog # PALBWN1201
Leverantör: Palbam
Gå till:
Product Details & Documents
When not in use this mini shelf can be folded down against the wall. Ideal for tight areas where space is limited. Rounded front edge.
- Fabrication material: Stainless steel 304 with electropolished finish
Specifikationer
- Beskrivning:Fold down mini shelf - solid
- B×D×H:1200×450×260 mm