Venting Screw N
Catalog # PAAR104284
Supplier: Anton Paar
New Product
Specifications
- Beskrivning:Venting screw, compatible with microwave reaction system, multiwave PRO
- Förp.:1
Specifications
About this item
Screw for safe and convenient closing and venting of pressure vessels NXF100 and NXQ80. Consists of a PEEK body with PTFE-TFM sealing bolt.