Workstation Spilltray and Drying Rack
Catalog # STUA67035-01
Leverantör: Cole Parmer
New Product
Specifikationer
- Beskrivning:Workstation spilltray and drying rack
- Förp.:1
- B×H:330,2×15,9 mm
Specifikationer
Om denna produkt
Reduce drying time and contain spilled liquids with this chemical resistant containment tray. Holds one ¹/₄ L of spilled liquid. Easy to clean simply lift off the white grid and empty.
- Chemical resistant
- Easy to clean
- Dishwasher safe
- Material: Polyethylene(PE)
- Length: 419 mm
Ordering information: One year warranty.