PELTOR™ HY80 Camelback Gel Sealing Rings
Katalog # MMMAHY80
Leverantör: 3M
New Product
Specifikationer
- Färg:Svart
- Beskrivning:Camelback gel sealing rings, HY80
- Material:PE
- Förp.:5
Specifikationer
Om denna produkt
Gel ear cushions for use on a wide variety of 3M™ PELTOR™ Headsets. These gel sealing rings features a super-thin polyurethane skin with a double-hump top face design and a silicon gel-filled bladder with a Confor™ foam backing.
- Super-soft foam for enhanced comfort
- Sealing rings feature a super-thin polyurethane skin with a double-hump top face design and a silicon gel-filled bladder with a Confor™ foam backing
Gel filled replacement cushions maintain a comfortable, noise-blocking seal around ears. Sealing rings maintain original service performance of Peltor products.