TASKI® Jontec Linosafe F1g Linoleum-Safe Stripper
Katalog # DIVY7512688
Leverantör: Diversey
New Product
Specifikationer
- Kapacitet:5 L
- Beskrivning:Linoleum safe stripper, 135W×185D×283H mm, 10,3 kg
- Förpackningstyp:Bottles
- Förp.:2 Can
Specifikationer
Om denna produkt
Linoleum safe stripper for removing emulsions and maintenance films from linoleum and other water-resistant hard floors. Prevents discolouring and damage to your Linoleum floor.
- Highly effective, slightly alkaline stripper
- Formulated for the safe removal of emulsions from linoleum floors
- Fact acting, low foaming
- Low odour
Benefits:
Quick and thorough removal of emulsion layers
Safe for use on all types of water-resistant hard floors, specially on Linoleum and calcium containing stones (example marble floors)
Easy and safe to use
Slightly perfumed for a pleasant use