Real Time PCR Detetction Test Kit Gluten
Katalog # BIOPGPCRSGLUTEN
Leverantör: BIOPREMIER
New Product
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:Real time PCR detection kit gluten, 100 reactions
Specifikationer
Om denna produkt
Gluten detection test kit by using qPCR. Celiac disease is an autoimmune disorder associated with an intolerance to gluten proteins mainly present in wheat, barley, and rye.
Methods are required to detect the presence of gluten contamination in food samples.