ContactUsLinkComponent
Bordet Gengou Agar Base, Dehydrated Culture Media
Bordet Gengou Agar Base, Dehydrated Culture Media
Catalog # HIMEM175-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
Bordet Gengou Agar Base, Dehydrated Culture Media
Catalog # HIMEM175-500G
Supplier:  HIMEDIA
New Product
CAS Number:  
Specifications
  • Förp.:
    1
  • Beskrivning:
    Bordet Gengou agar base, 500 g
Specifications
About this item
For the detection and isolation of Bordetella pertussis and Bordetella parapertussis.
  • Microbiological culture media
  • Packaged in 500 g poly bottles