ContactUsLinkComponent
Charcoal Agar Base Dehydrated Culture Media
  HIMEM344-500G
 :  HIMEDIA
New Product
 :  
Charcoal Agar Base Dehydrated Culture Media
  HIMEM344-500G
 :  HIMEDIA
New Product
 :  
  • Förp.:
    1
  • Beskrivning:
    Charcoal agar base, 500 g
For the cultivation of Bordetella pertussis, vaccine production, and stock culture maintenance.
  • Microbiological culture media
  • 500 g poly bottles