Wrench, universal, suited for Acros drums
Katalog # 473470010.
Leverantör: Thermo Fisher Scientific
New Product
Specifikationer
- Beskrivning:Wrench, universal, suited for Acros drums
- Längd:256 mm
- Material:Stål
- Use with head style:Open Sealed Caps and Drum Seals
- Förp.:1