VWR® 12-Channel Reagent Reservoirs
Supplier: VWR Collection
New Product
Certificates
About this item
These 12-channel reservoirs are made from high-quality, chemical resistant PP. Designed for serial dilutions or pipetting individual reagents using up to 12-channel pipetters.
V-shaped basins facilitate easy withdrawal of sample solutions
- Each channel holds up to 3 ml of solution and is individually numbered
- Includes a tight-fitting lid
- Individually wrapped packaging
- Non-sterile or sterilised by irradiation, SAL 10⁻⁶ (ISO 11137)
- Lot-certified, non-pyrogenic, DNase- and RNase-free
12-channel reagent reservoirs feature twelve 3 ml compartments, designed with V-shaped channels to maximise complete sample withdrawal. They are ideal for all liquid handling applications.
Certifications: ISO 13485 and ISO 9001.
Specifications
- Catalog No:
- 613-7300
- 613-7298
- 613-7309
- 613-7292
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Miljöcertificering:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M