Orders LinkContactUsLinkComponent
Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Human pluripotent stem cell-derived endoderm qPCR array, 1 KIT
Katalog # STMC07531
Marketsource
Leverantör:   Stemcell Technologies
Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Katalog # STMC07531
Leverantör:  Stemcell Technologies
Marketsource
CAS-nummer:  
Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Human pluripotent stem cell-derived endoderm qPCR array, 1 KIT
CAS-nummer:  
Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 1 KIT
Product Details & Documents
For characterisaton of hPSC-derived progenitor cells of definitive endoderm, hepatic, intestinal, or pancreatic lineages.
The Human Pluripotent Stem Cell (hPSC)-Derived Endoderm Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterisation of hPSC-derived definitive endodermal progenitor cells and their differentiated progeny including pancreatic, hepatic, and intestinal cells. This qPCR array provides the gene expression profile of hPSC-derived endodermal cells following in vitro differentiation, and can aid in development of protocols for directed differentiation towards endodermal lineages. Genes were selected based on their demonstrated expression in hPSCs, or in hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (D’Amour et al.), including pancreatic (Kroon et al.), hepatic (Hay et al.), and intestinal progenitor cells (Grapin-Botton & Melton).
qPCR is used to determine changes in steady-state mRNA levels of gene expression across multiple samples, generally normalised to the relative expression of internal control genes. Gene-specific primers amplify target sequences within cDNA pools reverse-transcribed from mRNA and, as with TaqMan® technology, hybridised sequence-specific probes provide a fluorescent signal from the 5' exonuclease activity of Taq DNA polymerase. The accumulation rate of the fluorescent signal is used to quantify the sample cDNA, and thereby the amount of mRNA in the original cell lysate.
This qPCR array contains validated primers and probes for detection of 90 genes whose expression is correlated with hPSCs or hPSC-derived ectodermal, mesodermal, and endodermal cells, or pancreatic, hepatic, and intestinal progenitor cells, as well as six endogenous (housekeeping) control genes. TBP (TATA box-binding protein) qPCR Control Template is provided as a synthetic DNA positive control.
Varning: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
Specifikationer
  • Förp.:
    1 KIT
  • Beskrivning:
    Human pluripotent stem cell-derived endoderm qPCR array