Individual Table for Suspended Cabinets
Katalog # KOTT505-00122
Leverantör: KOETTERMANN
New Product
Specifikationer
- Beskrivning:Individual mobile table with 'A' frame for suspended cabinets without board or shelf
- B×D×H:1254×700×870 mm
- Förp.:1
Specifikationer
Om denna produkt
The tables are fixed and mobile with an 'A' frame and depth of 700 mm.
The lower bar has been moved back, allowing the installation of suspended cabinets.
Ordering information: The board can be ordered separately.