AmMag™ Quatro Plasmid Purification Kit
Catalog # GENSL00882-24
Supplier: Genscript
New Product
CAS Number:
Specifications
- Förp.:1 KIT
- Beskrivning:Quatro plasmid purification kit, elution buffer: water, for 24 samples
Specifications
About this item
The AmMag Quatro is an automated solution for purification of large-scale, high quality, transfection-grade plasmids from Escherichia coli cultures. The solution consists of a system controller that provides simultaneous operation of up to 4 independent purification modules, along with pre-filled consumables and verified protocols.
- Kits are barcoded for lot tracking
- Individually packaged for single sample processing
- Enclosed design eliminates the possibility of cross-contamination
- Kit should be stored at 15 to 25 °C
The kit integrates unique endotoxin-removal methodology that consists of endotoxin washing solutions and endotoxin-removing column, while maintaining the stability of the supercoiled structure of the plasmid. The highly pure plasmid DNA can be directly used for downstream DNA plasmid workflows, such as enzyme digestion, PCR, sequencing, ligation, transformation and transfection.
Delivery: The AmMag Quatro consumable kit is an all-inclusive solution that contains Reagent cartridge - 24 cartridges (4×6); tip box - 24 boxes (4×6); sample tubes (50 ml centrifuge tube with lid) - 24 tubes; reaction tubes (50 ml centrifuge tube with lid) - 24 tubes, waste container (1).
Caution: Reagents in cartridge may precipitate when exposed to low temperatures (0 to 15 °C). Users can store the cartridge at room temperature for 24 hours or heat the cartridge in a water bath at 37 °C for 15 minutes, followed by shaking the cartridge to dissolve the precipitate.