Graduated Receiving Cylinder
Catalog # PAAR195234
Supplier: Anton Paar
New Product
Specifications
- Kapacitet:100 ml
- Beskrivning:Receiving cylinder with Viton O-ring, magnetic base
- Förp.:1
Specifications
About this item
Receiving cylinder with Viton O-ring,100 ml. Compatible with Diana 700, Diana 300.
Caution: Not suitable for solvents that dissolve rubber seals, e.g. acetone.