HIS Lite™ Cy5 Tris NTA Chelator
Catalog # AATB12659
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Förp.:100 µG
- Beskrivning:HIS Lite™ Cy5 Tris NTA chelator
Specifications
About this item
Cy5-Tris NTA compound is a sensitive fluorescent chelator for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces with the addition of certain metal ions (such as Ni²⁺, Co²⁺ etc.) as a mediator.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.