ABY Dye qPCR Calibration Solution *10000X*
Katalog # AATB67037
Leverantör: AAT BIOQUEST
New Product
CAS-nummer:
Specifikationer
- Förp.:100 µl
- Beskrivning:ABY Dye qPCR calibration solution *10000X*
Specifikationer
Om denna produkt
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.