ContactUsLinkComponent
ReadiLink™ Rapid XFD750 Antibody Labelling Kit
Catalog # AATB5745
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
ReadiLink™ Rapid XFD750 Antibody Labelling Kit
Catalog # AATB5745
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Förp.:
    3
  • Pk Info:
    3×1 mg
Specifications
About this item
ReadiLink™ Rapid antibody labelling kits, designed for production scale, provide a convenient and efficient method for labelling large volumes of antibodies with XFD dyes (equivalent to Alexa Fluor®).
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Delivery: Shipped at RT.
Caution: Research use only.