ReadiLink™ Rapid mFluor™ Violet 610 Antibody Labelling Kit
Katalog # AATB1116
Leverantör: AAT BIOQUEST
New Product
CAS-nummer:
Specifikationer
- Förp.:2
- Includes:2 labellings
Specifikationer
Om denna produkt
Microscale optimised for labeling 50 µg antibody per reaction. ReadiLink™ Rapid mFluor™ Violet 610 antibody labelling kits provide a quick and convenient method to label antibodies, proteins (<10 kDa), or other amine-containing biomolecules.
- Robust protocol
- Consistent results
- 12-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry and microscopy.
Delivery: Shipped at RT.
Caution: Research use only.