HIS Lite™ OG488-Tris NTA Chelator
Catalog # AATB12655
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Förp.:100 µG
- Beskrivning:HIS Lite™ OG488-tris NTA chelator
Specifications
About this item
HIS Lite™ OG488-Tris NTA Chelator is used as a sensitive green fluorescent probe for detecting polyhistidine-labelled proteins in combination with the addition of a heavy metal ion such as Ni²⁺ in cells, solution and solid surfaces.
- Stable for 24 months
Quality tested in-house.
Caution: For research use only.