FAM Dye qPCR Calibration Solution *10000X*
Catalog # AATB67032
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Förp.:100 µl
- Beskrivning:FAM dye qPCR calibration solution *10000X*
Specifications
About this item
FAM dye qPCR calibration solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ seven systems.
- Stable for 12 months
Calibration and verification should be run every six months or as needed.
Caution: For research use only.