Supelco® TraceCERT® Organic Reference Standard, Pinoxaden Metabolite SYN502836
CRM80876-10MG
: Merck
:
- Förp.:10 mg
- Standard:Certified reference material, TraceCERT®
- CAS-nr:881376-42-7
- Beskrivning:Pinoxaden metabolite SYN502836
- Storage conditions:2…8 °C
- Faroklassbeskrivning:H302 + H332 - H319 - H334 - H335 - H361d - H410
- Shipping conditions:WICE
- M-Clarity quality level:MQ300
- Synonym(s):3,5-Diethyl-4-(hexahydro-7,9-dioxo-7H-pyrazolo[1,2-d][1,4,5]oxadiazepin-8-yl)-benzoic acid
- Molekylvikt:346,38
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P201 - P273 - P301 + P312 - P304 + P340 + P312 - P305 + P351 + P338 - P308 + P313