Order Entry
Sweden
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Tritosulfurone Metabolite M635H004
Supelco® TraceCERT® Organic Reference Standard, Tritosulfurone Metabolite M635H004
Katalog # CRM80839-10MG
Leverantör:  Merck
CAS-nummer:  
Supelco® TraceCERT® Organic Reference Standard, Tritosulfurone Metabolite M635H004
Katalog # CRM80839-10MG
Leverantör:  Merck
CAS-nummer:  

Specifikationer

  • Förp.:
    10 mg
  • Standard:
    Certified reference material, TraceCERT®
  • CAS-nr:
    5311-05-7
  • Beskrivning:
    Tritosulfurone metabolite M635H004
  • Storage conditions:
    2…8 °C
  • Faroklassbeskrivning:
    H302 - H361d
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01679229
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    2-Amino-4-methoxy-6-(trifluoromethyl)-1,3,5-triazine
  • Molekylvikt:
    194,11

Specifikationer

Om denna produkt

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P201 - P202 - P264 - P270 - P301 + P312 - P308 + P313