Order Entry
Sweden
ContactUsLinkComponent
Anti-RPL27A Rabbit Polyclonal Antibody
Anti-RPL27A Rabbit Polyclonal Antibody
Katalog # ANTIA95730-100
Leverantör:  ANTIBODIES.COM
New Product
undefined
Anti-RPL27A Rabbit Polyclonal Antibody
Katalog # ANTIA95730-100
Leverantör:  ANTIBODIES.COM
New Product

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    60S ribosomal protein L27a
  • Antigensymbol:
    RPL27A
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# P46776
  • Antigen synonymer:
    RPL27A|FLJ43464|RL27A_HUMAN
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molekylvikt:
    16 kDa
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human RPL27A (amino acids 99-148)
  • Rening:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Rabbit polyclonal antibody to RPL27A for WB, IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IF, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000

Type: Primary
Antigen: RPL27A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat