Anti-Calbindin Mouse Recombinant Antibody [clone: rCALB1/8499]
Katalog # ANTIA314535-100
Leverantör: ANTIBODIES.COM
New Product
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:avian-type
- Antigensymbol:Calbindin
- Klonalitet:Recombinant
- Klon:rCALB1/8499
- Konjugation:Unconjugated
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgG1 kappa
- Reaktivitet:Human
- Form:Liquid
- Gen-ID:UniprotID# P05937
- Antigen synonymer:Calbindin|Calbindin D28|CALB|CALB1_HUMAN|CALB1|D 28K|Calbindin 1 28kDa|CALB 1|CAB27
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molekylvikt:52 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 7-96) of human CALB1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Rening:Protein A/G chromatography
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifikationer
Om denna produkt
Recombinant mouse monoclonal [rCALB1/8499] antibody to Calbindin for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Calbindin
Clonality: Recombinant
Clone: rCALB1/8499
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human