Anti-IgG Goat Polyclonal Antibody (trFluor™ Tb)
Leverantör: AAT BIOQUEST
New Product
Certifikat
Om denna produkt
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Purified and quality-tested for consistent results
- Suitable for multiple applications
- Purified through affinity chromatography
This trFluor™ Tb goat anti-rabbit IgG (H+L) conjugate is commonly used as a second step reagent for indirect immunofluorescent staining, when used in conjunction with primary antibodies.
Caution: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: trFluor™ Tb
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Specifikationer
- Catalog No:
- AATB16848
- AATB16726
- Antigensymbol:
- IgG
- IgG
- Antikroppsnamn:
- Immunoglobulin G
- Immunoglobulin G
- Antikroppstyp:
- Sekundär
- Sekundär
- Klonalitet:
- Polyclonal
- Polyclonal
- Konjugation:
- trFluor™ Tb
- trFluor™ Tb
- Reaktivitet:
- Rabbit
- Rabbit
- Värd:
- Goat
- Goat
- Isotyp:
- IgG (H, L)
- IgG (H, L)
- Rening:
- Affinity chromatography
- Affinity chromatography
- Korsad adsorption:
- Minimal cross reaction to human, horse, rabbit and bovine
- Minimal cross reaction to human, horse, rabbit and bovine
- Förvaringstemperatur:
- Freeze (<−15 °C); Minimise light exposure
- Freeze (<−15 °C); Minimise light exposure
- Shipping temperature:
- 20
- 20