Anti-Podocalyxin Rabbit Recombinant Antibody [clone: PDPN/4009R]
Catalog # ANTIA251332-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:GCTM-2 antigen
- Antigensymbol:Podocalyxin
- Klonalitet:Recombinant
- Klon:PDPN/4009R
- Konjugation:Unconjugated
- ELISA:Yes
- Värd:Rabbit
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG
- Reaktivitet:Human
- Form:Liquid
- Gen-ID:UniprotID# Q86YL7
- Antigen synonymer:PCLP|Podocalyxin|PCLP-1|MGC138240|PCLP1|Gp200|PC|Gp2|Pcx
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molekylvikt:17 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Koncentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to amino acids within 1 - 100 of Podoplanin
- Tested applications:IHC-P
- Rening:Protein A/G chromatography
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [PDPN/4009R] antibody to Podocalyxin for ELISA, IF and IHC-P with samples derived from Human.
- Validated applications: ELISA, IF, IHC-P
- Recommended dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Podocalyxin
Clonality: Recombinant
Clone: PDPN/4009R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human