Order Entry
Sweden
ContactUsLinkComponent
 
Anti-MerTK Mouse Monoclonal Antibody [clone: MERTK/3022]
  ANTIA251301-100
New Product
undefined
Anti-MerTK Mouse Monoclonal Antibody [clone: MERTK/3022]
  ANTIA251301-100
New Product

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    c MER
  • Antigensymbol:
    MerTK
  • Klonalitet:
    Monoclonal
  • Klon:
    MERTK/3022
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG2a kappa
  • Reaktivitet:
    Human
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# Q12866
  • Antigen synonymer:
    MERPEN|MERTK|c-mer|MERK|cmer protooncogene tyrosine kinase|c mer proto oncogene tyrosine kinase|MER receptor tyrosine kinase|Eyk|cMER
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molekylvikt:
    110 kDa
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Koncentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 55 - 148, of human MERTK protein. The exact sequence is proprietary
  • Tested applications:
    IHC-P
  • Rening:
    Protein A/G chromatography
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

 

 

Mouse monoclonal [MERTK/3022] antibody to MerTK for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: MerTK
Clonality: Monoclonal
Clone: MERTK/3022
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human