Anti-Glypican 3 Mouse Recombinant Antibody [clone: rGPC3/863]
ANTIA251913-100
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:DGSX
- Antigensymbol:Glypican 3
- Klonalitet:Recombinant
- Klon:rGPC3/863
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Mouse
- ImmunKemi:Yes
- ImmunoFluorescens:Yes
- Isotyp:IgG1 kappa
- Reaktivitet:Human,Rat
- Form:Liquid
- Gen-ID:UniprotID# P51654
- Antigen synonymer:Gpc3|Heparan sulphate proteoglycan|Glypican 3|Glypican-3 [Precursor]|Intestinal protein OCI 5|GTR2-2|GTR2 2|GPC3_HUMAN|Glypican proteoglycan 3
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molekylvikt:67 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human GPC3 protein
- Tested applications:IHC-P
- Rening:Protein A/G chromatography
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Recombinant mouse monoclonal [rGPC3/863] antibody to Glypican 3 for Flow Cytometry, IF and IHC-P with samples derived from Human and Rat.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Glypican 3
Clonality: Recombinant
Clone: rGPC3/863
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat