Anti-Growth Hormone Mouse Recombinant Antibody [clone: rGH/1450]
Katalog # ANTIA248716-100
Leverantör: ANTIBODIES.COM
New Product
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:gH
- Antigensymbol:Growth Hormone
- Klonalitet:Recombinant
- Klon:rGH/1450
- Konjugation:Unconjugated
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgG1 kappa
- Reaktivitet:Human
- Form:Liquid
- Gen-ID:UniprotID# P01241
- Antigen synonymer:GH-N|Growth hormone, normal|Growth hormone 1|Growth Hormone|GHB5|Growth hormone B5|GHN|GH1|Growth hormone, pituitary
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molekylvikt:20 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, within amino acids 58-187, of human Growth Hormone. The exact sequence is proprietary.
- Tested applications:IHC-P
- Rening:Protein A/G chromatography
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifikationer
Om denna produkt
Recombinant mouse monoclonal [rGH/1450] antibody to Growth Hormone for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Growth Hormone
Clonality: Recombinant
Clone: rGH/1450
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human