Order Entry
Sweden
ContactUsLinkComponent
 
Anti-MTHFR Goat Polyclonal Antibody
  ANTIA286013-100
New Product
Anti-MTHFR Goat Polyclonal Antibody
  ANTIA286013-100
New Product

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    5 10 methylenetetrahydrofolate reductase
  • Antigensymbol:
    MTHFR
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Goat
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Form:
    Liquid
  • Gen-ID:
    4524
  • Antigen synonymer:
    MTHFR|Methylenetetrahydrofolate reductase intermediate form|MTHR_HUMAN|5 10 methylenetetrahydrofolate reductase (NADPH)|Methylenetetrahydrofolate reductase (NAD(P)H)
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,3, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-NRPTQNARETEAP
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Koncentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to the C terminus of human MTHFR.
  • Tested applications:
    IHC
  • Rening:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

 

 

Goat polyclonal antibody to MTHFR for ELISA and IHC with samples derived from human.

  • Validated applications: ELISA, IHC

Type: Primary
Antigen: MTHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human