- Förp.:96 Tests
- Analysens varaktighet:One step
- Analystyp:Sandwich
- Formatera:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Ugcg
- Beskrivning:Mouse Ugcg ELISA kit
- Typ av prov:Serum, plasma or other biological fluids
- Detektionsmetod:Colorimetric
- Time to Results:1 h 30 min
- Hållbarhetstid:Store for 6 months at 4 °C
- Detektionsområde:25 - 400 ng/L
- Förvaringstemperatur:4 °C
- Provvolym:40 μl
- Känslighet:0,96 ng/L
- Regulatory status:RUO
Mouse Ugcg ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Ugcg in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 25 to 400 ng/L
- Sensitivity: 0,96 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse Ugcg ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Ugcg in An antibody specific for Ugcg has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Ugcg present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Ugcg antibody, which also binds the Ugcg present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Ugcg antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Ugcg antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Ugcg captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Ugcg in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.