Order Entry
Sweden
ContactUsLinkComponent
Human NOSTRIN ELISA Kit
Human NOSTRIN ELISA Kit
Katalog # ANTIA247316-96
Leverantör:  ANTIBODIES.COM
New Product
CAS-nummer:  
undefined
Human NOSTRIN ELISA Kit
Katalog # ANTIA247316-96
Leverantör:  ANTIBODIES.COM
New Product
CAS-nummer:  

Specifikationer

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    NOSTRIN
  • Beskrivning:
    Human NOSTRIN ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Korsreaktivitet:
    Human NOSTRIN ELISA Kit exhibits high specificity and excellent specificity for the detection of human NOSTRIN. No significant cross-reactivity or interference between NOSTRIN and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    0,313 - 20 ng/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    0,188 ng/ml
  • Regulatory status:
    RUO

Specifikationer

Om denna produkt

Human NOSTRIN ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human NOSTRIN in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human NOSTRIN ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human NOSTRIN in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for NOSTRIN has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the NOSTRIN present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-NOSTRIN Antibody, which binds the captured NOSTRIN present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of NOSTRIN captured in each well. The concentration of NOSTRIN can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.