Order Entry
Sweden
ContactUsLinkComponent
Rabbit iNOS ELISA Kit
Rabbit iNOS ELISA Kit
Catalog # ANTIA73939-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rabbit iNOS ELISA Kit
Catalog # ANTIA73939-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    iNOS
  • Beskrivning:
    Rabbit iNOS ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Rabbit iNOS ELISA Kit exhibits high specificity and excellent specificity for the detection of rabbit iNOS. No significant cross-reactivity or interference between iNOS and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    0,313 - 20 ng/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    0,188 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rabbit iNOS ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rabbit iNOS in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Detection Range: 0,313 to 20 ng/ml
  • Sensitivity: 0,188 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rabbit iNOS ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rabbit iNOS in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for iNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the iNOS present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-iNOS Antibody, which binds the captured iNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of iNOS captured in each well. The concentration of iNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.