Order Entry
Sweden
ContactUsLinkComponent
Rat Liver FABP ELISA Kit
Rat Liver FABP ELISA Kit
Katalog # ANTIA79315-96
Leverantör:  ANTIBODIES.COM
New Product
CAS-nummer:  
undefined
Rat Liver FABP ELISA Kit
Katalog # ANTIA79315-96
Leverantör:  ANTIBODIES.COM
New Product
CAS-nummer:  

Specifikationer

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Liver FABP
  • Beskrivning:
    Rat liver FABP ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Rat Liver FABP ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Liver FABP. No significant cross-reactivity or interference between Liver FABP and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    1,563 - 100 ng/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    0,938 ng/ml
  • Regulatory status:
    RUO

Specifikationer

Om denna produkt

Rat Liver FABP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Liver FABP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 1,563 to 100 ng/ml
  • Sensitivity: 0,938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Liver FABP ELISA kit (A79315) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Liver FABP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Liver FABP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Liver FABP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Liver FABP Antibody, which binds the captured Liver FABP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Liver FABP captured in each well. The concentration of Liver FABP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.