Order Entry
Sweden
ContactUsLinkComponent
Rat ZO1 Tight Junction Protein ELISA Kit
Rat ZO1 Tight Junction Protein ELISA Kit
Catalog # ANTIA79748-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Rat ZO1 Tight Junction Protein ELISA Kit
Catalog # ANTIA79748-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    ZO1 Tight Junction Protein
  • Beskrivning:
    Rat ZO1 tight junction protein ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Rat ZO1 Tight Junction Protein ELISA Kit exhibits high specificity and excellent specificity for the detection of rat ZO1 Tight Junction Protein. No significant cross-reactivity or interference between ZO1 Tight Junction Protein and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    0,156 - 10 ng/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    0,094 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Rat ZO1 Tight Junction Protein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ZO1 Tight Junction Protein in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat ZO1 Tight Junction Protein ELISA kit (A79748) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat ZO1 Tight Junction Protein in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for ZO1 Tight Junction Protein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ZO1 Tight Junction Protein present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-ZO1 Tight Junction Protein Antibody, which binds the captured ZO1 Tight Junction Protein present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of ZO1 Tight Junction Protein captured in each well. The concentration of ZO1 Tight Junction Protein can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.