Order Entry
Sweden
ContactUsLinkComponent
Human D Amino Acid Oxidase ELISA Kit
Human D Amino Acid Oxidase ELISA Kit
Catalog # ANTIA79255-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human D Amino Acid Oxidase ELISA Kit
Catalog # ANTIA79255-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    D Amino Acid Oxidase
  • Beskrivning:
    Human D amino acid oxidase ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Human D Amino Acid Oxidase ELISA kit exhibits high specificity and excellent specificity for the detection of human D Amino Acid Oxidase. No significant cross-reactivity or interference between D Amino Acid Oxidase and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    0,781 - 50 ng/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    0,469 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human D Amino Acid Oxidase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human D Amino Acid Oxidase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0,781 - 50 ng/ml
  • Sensitivity: 0,469 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human D Amino Acid Oxidase ELISA kit (A79255) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human D Amino Acid Oxidase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for D Amino Acid Oxidase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the D Amino Acid Oxidase present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-D Amino Acid Oxidase Antibody, which binds the captured D Amino Acid Oxidase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of D Amino Acid Oxidase captured in each well. The concentration of D Amino Acid Oxidase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.