Specifications
- Förp.:96 Tests
- Analysens varaktighet:Multiple steps
- Analystyp:Sandwich
- Formatera:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Prokineticin receptor 1
- Beskrivning:Rat Prokineticin receptor 1 ELISA kit
- Typ av prov:Serum, plasma, tissue homogenates and other biological fluids
- Korsreaktivitet:Rat Prokineticin receptor 1 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Prokineticin receptor 1. No significant cross-reactivity or interference between Prokineticin receptor 1 and analogues was observed.
- Detektionsmetod:Colorimetric
- Time to Results:4 h 30 min
- Analysprincip:Quantitative
- Hållbarhetstid:Store for 6 months at 4 °C
- Detektionsområde:0,156 - 10 ng/ml
- Förvaringstemperatur:4 °C
- Provvolym:100 μl
- Känslighet:0,094 ng/ml
- Regulatory status:RUO
Specifications
About this item
Rat Prokineticin receptor 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Prokineticin receptor 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 0,156 to 10 ng/ml
- Sensitivity: 0,094 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Rat Prokineticin Receptor 1 ELISA Kit (A79642) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Prokineticin Receptor 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Prokineticin Receptor 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Prokineticin Receptor 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Prokineticin Receptor 1 Antibody, which binds the captured Prokineticin Receptor 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Prokineticin Receptor 1 captured in each well. The concentration of Prokineticin Receptor 1 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.