Specifikationer
- Förp.:96 Tests
- Analysens varaktighet:Multiple steps
- Analystyp:Sandwich
- Formatera:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Wilms Tumor Protein
- Beskrivning:Human Wilms Tumor protein ELISA kit
- Typ av prov:Serum, plasma, tissue homogenates and other biological fluids
- Korsreaktivitet:Human Wilms Tumor Protein ELISA Kit exhibits high specificity and excellent specificity for the detection of human Wilms Tumor Protein. No significant cross-reactivity or interference between Wilms Tumor Protein and analogues was observed.
- Detektionsmetod:Colorimetric
- Time to Results:4 h 30 min
- Analysprincip:Quantitative
- Hållbarhetstid:Store for 6 months at 4 °C
- Detektionsområde:15,625 - 1000 pg/ml
- Förvaringstemperatur:4 °C
- Provvolym:100 μl
- Känslighet:9,375 pg/ml
- Regulatory status:RUO
Specifikationer
Om denna produkt
Human Wilms Tumor Protein ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Wilms Tumor Protein in serum, plasma, tissue homogenates, and other biological fluids.
- Ready To Use ELISA Kit
- Detection Range: 15,625 to 1000 pg/ml
- Sensitivity: 9,375 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%
Human Wilms Tumor Protein ELISA Kit (A79797) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Wilms Tumor Protein in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Wilms Tumor Protein has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Wilms Tumor Protein present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Wilms Tumor Protein Antibody, which binds the captured Wilms Tumor Protein present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Wilms Tumor Protein captured in each well. The concentration of Wilms Tumor Protein can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.