Order Entry
Sweden
ContactUsLinkComponent
 
Human Dopamine beta Hydroxylase ELISA Kit
Katalog # ANTIA79257-96
Leverantör:  ANTIBODIES.COM
New Product
CAS-nummer:  
undefined
Human Dopamine beta Hydroxylase ELISA Kit
Katalog # ANTIA79257-96
Leverantör:  ANTIBODIES.COM
New Product
CAS-nummer:  

Specifikationer

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Dopamine beta Hydroxylase
  • Beskrivning:
    Human dopamine beta hydroxylase ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Human Dopamine beta Hydroxylase ELISA Kit exhibits high specificity and excellent specificity for the detection of human Dopamine beta Hydroxylase. No significant cross-reactivity or interference between Dopamine beta Hydroxylase and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    1,563 - 100 ng/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    0,938 ng/ml
  • Regulatory status:
    RUO

Specifikationer

Om denna produkt

Human Dopamine beta Hydroxylase ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Dopamine beta Hydroxylase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 1,563 to 100 ng/ml
  • Sensitivity: 0,938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Dopamine beta Hydroxylase ELISA kit (A79257) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Dopamine beta Hydroxylase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Dopamine beta Hydroxylase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Dopamine beta Hydroxylase present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Dopamine beta Hydroxylase Antibody, which binds the captured Dopamine beta Hydroxylase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Dopamine beta Hydroxylase captured in each well. The concentration of Dopamine beta Hydroxylase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.