Specifikationer
- Förp.:96 Tests
- Analysens varaktighet:Multiple steps
- Analystyp:Sandwich
- Formatera:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Transferrin Receptor 1
- Beskrivning:Mouse transferrin receptor 1 ELISA kit
- Typ av prov:Serum, plasma, tissue homogenates and other biological fluids
- Korsreaktivitet:Mouse Transferrin Receptor 1 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse Transferrin Receptor 1. No significant cross-reactivity or interference between Transferrin Receptor 1 and analogues was observed.
- Detektionsmetod:Colorimetric
- Time to Results:4 h 30 min
- Analysprincip:Quantitative
- Hållbarhetstid:Store for 6 months at 4 °C
- Detektionsområde:0,625 - 40 ng/ml
- Förvaringstemperatur:4 °C
- Provvolym:100 μl
- Känslighet:0,375 ng/ml
- Regulatory status:RUO
Specifikationer
Om denna produkt
Mouse Transferrin Receptor 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Transferrin Receptor 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0,625 - 40 ng/ml
- Sensitivity: 0,375 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse Transferrin Receptor 1 ELISA kit (A78864) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Transferrin Receptor 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Transferrin Receptor 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Transferrin Receptor 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Transferrin Receptor 1 Antibody, which binds the captured Transferrin Receptor 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Transferrin Receptor 1 captured in each well. The concentration of Transferrin Receptor 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.