Order Entry
Sweden
ContactUsLinkComponent
Human Peptide YY/PYY ELISA Kit
Human Peptide YY/PYY ELISA Kit
Catalog # ANTIA79680-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Peptide YY/PYY ELISA Kit
Catalog # ANTIA79680-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Competitive
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Peptide YY/PYY
  • Beskrivning:
    Human peptide YY/PYY ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Human Peptide YY/PYY ELISA Kit exhibits high specificity and excellent specificity for the detection of human Peptide YY/PYY. No significant cross-reactivity or interference between Peptide YY/PYY and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    31,25 - 2000 pg/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    50 μl
  • Känslighet:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Peptide YY/PYY ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human Peptide YY/PYY in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Peptide YY/PYY ELISA kit (A79680) employs the competitive enzyme immunoassay technique for the quantitative measurement of human Peptide YY/PYY in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Peptide YY/PYY antigen. During the incubation, Peptide YY/PYY present in the samples or standards competes with the fixed amount of immobilized Peptide YY/PYY for binding sites on the Biotinylated Anti-Peptide YY/PYY Antibody. The more Peptide YY/PYY present in a sample or standard, the less Biotinylated Anti-Peptide YY/PYY Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Peptide YY/PYY Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Peptide YY/PYY present in each sample or standard. The concentration of Peptide YY/PYY can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.