Specifikationer
- Förp.:96 Tests
- Analysens varaktighet:Multiple steps
- Analystyp:Sandwich
- Formatera:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Lipoteichoic Acids
- Beskrivning:Human lipoteichoic acids ELISA kit
- Typ av prov:Serum, plasma, tissue homogenates and other biological fluids
- Korsreaktivitet:Human Lipoteichoic Acids ELISA kit exhibits high specificity and excellent specificity for the detection of human Lipoteichoic Acids. No significant cross-reactivity or interference between Lipoteichoic Acids and analogues was observed.
- Detektionsmetod:Colorimetric
- Time to Results:4 h 30 min
- Analysprincip:Quantitative
- Hållbarhetstid:Store for 6 months at 4 °C
- Detektionsområde:7,813 - 500 pg/ml
- Förvaringstemperatur:4 °C
- Provvolym:100 μl
- Känslighet:4,688 pg/ml
- Regulatory status:RUO
Specifikationer
Om denna produkt
Human Lipoteichoic Acids ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Lipoteichoic Acids in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 7,813 - 500 pg/ml
- Sensitivity: 4,688 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human Lipoteichoic Acids ELISA kit (A79811) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Lipoteichoic Acids in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Lipoteichoic Acids has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Lipoteichoic Acids present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Lipoteichoic Acids Antibody, which binds the captured Lipoteichoic Acids present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Lipoteichoic Acids captured in each well. The concentration of Lipoteichoic Acids can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.