Order Entry
Sweden
ContactUsLinkComponent
Human Cathepsin F ELISA Kit
Human Cathepsin F ELISA Kit
Catalog # ANTIA77938-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human Cathepsin F ELISA Kit
Catalog # ANTIA77938-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Förp.:
    96 Tests
  • Analysens varaktighet:
    Multiple steps
  • Analystyp:
    Sandwich
  • Formatera:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Cathepsin F
  • Beskrivning:
    Human Cathepsin F ELISA kit
  • Typ av prov:
    Serum, plasma, tissue homogenates and other biological fluids
  • Korsreaktivitet:
    Human Cathepsin F ELISA kit exhibits high specificity and excellent specificity for the detection of human Cathepsin F. No significant cross-reactivity or interference between Cathepsin F and analogues was observed.
  • Detektionsmetod:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Analysprincip:
    Quantitative
  • Hållbarhetstid:
    Store for 6 months at 4 °C
  • Detektionsområde:
    78,125 - 5000 pg/ml
  • Förvaringstemperatur:
    4 °C
  • Provvolym:
    100 μl
  • Känslighet:
    46,875 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human Cathepsin F ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cathepsin F in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Cathepsin F ELISA kit (A77938) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Cathepsin F in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cathepsin F has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cathepsin F present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cathepsin F Antibody, which binds the captured Cathepsin F present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cathepsin F captured in each well. The concentration of Cathepsin F can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.