Centering Ring, KF25SVCR
Katalog # EDWAKF25SVCR
Leverantör: EDWARDS HIGH VACUUM METTLER
New Product
Specifikationer
- Beskrivning:Center ring used for vacuum hose connection to rotary pump
- Förp.:1
Specifikationer
Om denna produkt
Used for vacuum hose connection to rotary pump.
- Stainless steel with a black o-ring
- Compatiable with all Edwards and Leybold pump options